5-Amino-4-bromo-2-fluorobenzonitrile 98%

Supplier: Ambeed
Warning

A375799-100MG A375799-10G A375799-1G A375799-250MG
76828-930EA 15 USD
76828-930 76828-932 76828-934 76828-936
5-Amino-4-bromo-2-fluorobenzonitrile 98%
5-Amino-4-bromo-2-fluorobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₄BrFN₂
MDL Number: MFCD11977386
CAS Number: 893615-28-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR