2-Chloro-5-propoxyphenylboronic acid 96%

Supplier: Ambeed
Warning

Synonyms: (2-Chloro-5-propoxyphenyl)boronic acid

A373906-1G A373906-250MG
76828-918EA 128.37 USD
76828-918 76828-920
2-Chloro-5-propoxyphenylboronic acid 96%
2-Chloro-5-propoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₂BClO₃
MW: 214.4538 g/mol
MDL Number: MFCD16295074
CAS Number: 1256346-14-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR