4-(Butylsulfonyl)phenylboronic acid 96%

Supplier: Ambeed
Warning

Synonyms: (4-(Butylsulphonyl)phenyl)boronic acid

A373856-100MG A373856-1G A373856-250MG
76828-912EA 33.95 USD
76828-912 76828-914 76828-916
4-(Butylsulfonyl)phenylboronic acid 96%
4-(Butylsulphonyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₁₁BO₄S
MW: 242.1000 g/mol
MDL Number: MFCD12546549
CAS Number: 1217501-02-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR