Methyl 6-bromo-3-hydroxypicolinate 98%

Supplier: Ambeed
Warning

A372972-100MG A372972-250MG
76828-910EA 45.62 USD
76828-910 77613-112
Methyl 6-bromo-3-hydroxypicolinate 98%
Methyl 6-bromo-3-hydroxypicolinate

Precaución: for Research Use Only

Formula: C₇H₆BrNO₃
MW: 232.03 g/mol
MDL Number: MFCD12828055
CAS Number: 321601-48-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR