2-Chloro-6-(ethylamino)-4-nitrophenol 95%

Supplier: Ambeed
Danger

A372562-1G A372562-250MG A372562-25G A372562-5G
76828-894EA 49.87 USD
76828-894 76828-896 76828-898 76828-900
2-Chloro-6-(ethylamino)-4-nitrophenol 95%
2-Chloro-6-(ethylamino)-4-nitrophenol

Precaución: for Research Use Only

Formula: C₈H₉ClN₂O₃
MW: 216.62 g/mol
MDL Number: MFCD11111995
CAS Number: 131657-78-8
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR