2-Cyano-4-fluorophenylboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 2-Cyano-4-fluorophenylboronic Acid Pinacol Ester, 2-Cyano-4-fluorobenzeneboronic acid pinacol ester

A371003-1G A371003-250MG A371003-5G
76828-870EA 27 USD
76828-870 76828-872 76828-874
2-Cyano-4-fluorophenylboronic acid pinacol ester 98%
2-Cyano-4-fluorophenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₅BFNO₂
MDL Number: MFCD09260444
CAS Number: 461451-63-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR