3,6-Diphenyl-2,5-dihydropyrrolo[3,4-c]pyrrole-1,4-dione 98%

Supplier: Ambeed
Warning

Synonyms: 3,6-Diphenylpyrrolo[3,4-c]pyrrole-1,4(2H,5H)-dione, Pigment Red 255

A369089-1G A369089-5G
76828-848EA 21.22 USD
76828-848 76828-850
3,6-Diphenyl-2,5-dihydropyrrolo[3,4-c]pyrrole-1,4-dione 98%
3,6-Diphenyl-2,5-dihydropyrrolo[3,4-c]pyrrole-1,4-dione

Precaución: for Research Use Only

Formula: C₁₈H₁₂N₂O₂
MW: 288.31 g/mol
MDL Number: MFCD11870113
CAS Number: 54660-00-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR