Benzo[b]thiophene-3-carbonitrile 98%
Supplier: Ambeed
|
Warning
|
A369014-1G
A369014-250MG
A369014-5G
76828-842EA
46.68
USD
76828-842
76828-844
76828-846
Benzo[b]thiophene-3-carbonitrile 98%
Benzo[b]thiophene-3-carbonitrile
Precaución: for Research Use Only
Formula:
C₉H₅NS MW: 159.208 g/mol |
MDL Number:
MFCD08276872 CAS Number: 24434-84-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...