N-Methoxy-N-methylbutyramide 98%

Supplier: Ambeed
Warning

A367139-1G A367139-5G
76828-814EA 17 USD
76828-814 76828-816
N-Methoxy-N-methylbutyramide 98%
N-Methoxy-N-methylbutyramide

Precaución: for Research Use Only

Formula: C₆H₁₃NO₂
MDL Number: MFCD14707556
CAS Number: 109480-78-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR