2-Cyano-3-nitrobenzoic acid 97%

Supplier: Ambeed
Warning

A366204-10G A366204-1G A366204-250MG A366204-5G
76828-806EA 356 USD
76828-806 76828-808 76828-810 76828-812
2-Cyano-3-nitrobenzoic acid 97%
2-Cyano-3-nitrobenzoic acid

Precaución: for Research Use Only

Formula: C₈H₄N₂O₄
MDL Number: MFCD09745182
CAS Number: 1260834-31-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR