4-Carbamoyl-3-chlorobenzeneboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (4-Carbamoyl-3-chlorophenyl)boronic acid, 4-(Aminocarbonyl)-3-chlorobenzeneboronic acid, 4-Aminocarbonyl-3-chlorophenylboronic acid

A365815-1G A365815-250MG A365815-5G
76828-800EA 80 USD
76828-800 76828-802 76828-804
4-Carbamoyl-3-chlorobenzeneboronic acid 98%
4-Carbamoyl-3-chlorobenzeneboronic acid

Precaución: for Research Use Only

Formula: C₇H₇BClNO₃
MDL Number: MFCD07363777
CAS Number: 850589-52-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR