4'-Pentyloxybiphenyl-4-boronic acid 95%

Supplier: Ambeed
Warning

Synonyms: [4'-(Pentyloxy)-[1,1'-biphenyl]-4-yl]boronic acid, (4'-(Pentyloxy)-[1,1'-biphenyl]-4-yl)boronic acid, 4'-Amyloxybiphenyl-4-boronic acid

A364638-10G A364638-1G A364638-5G
76828-794EA 44 USD
76828-794 76828-796 76828-798
4'-Pentyloxybiphenyl-4-boronic acid 95%
4'-Pentyloxybiphenyl-4-boronic acid

Precaución: for Research Use Only

Formula: C₁₇H₂₁BO₃
MW: 284.16 g/mol
MDL Number: MFCD07644475
CAS Number: 158937-25-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR