4-Bromoquinoline-3-carboxylic acid ethyl ester 95%

Supplier: Ambeed
Warning

Synonyms: Ethyl 4-bromoquinoline-3-carboxylate

A364158-1G A364158-250MG A364158-5G
76828-784EA 50.93 USD
76828-784 76828-786 76828-788
4-Bromoquinoline-3-carboxylic acid ethyl ester 95%
4-Bromoquinoline-3-carboxylic acid ethyl ester

Precaución: for Research Use Only

Formula: C₁₂H₁₀BrNO₂
MW: 280.117 g/mol
MDL Number: MFCD16987479
CAS Number: 1242260-12-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR