4-(2,2,2-Trifluoroethylcarbamoyl)phenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (4-((2,2,2-Trifluoroethyl)carbamoyl)phenyl)boronic acid

A364049-100MG A364049-1G A364049-250MG A364049-5G
76828-776EA 28 USD
76828-776 76828-778 76828-780 76828-782
4-(2,2,2-Trifluoroethylcarbamoyl)phenylboronic acid 98%
4-(2,2,2-Trifluoroethylcarbamoyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₉H₉BF₃NO₃
MDL Number: MFCD08235087
CAS Number: 874459-90-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR