N-Isopropylmethacrylamide 97%

Supplier: Ambeed
Warning

Synonyms: 2-methyl-N-(propan-2-yl)prop-2-enamide, N-isopropyl-2-methyl-prop-2-enamide

A363187-1G A363187-5G
76828-770EA 5.31 USD
76828-770 76828-772
N-Isopropylmethacrylamide 97%
N-Isopropylmethacrylamide

Precaución: for Research Use Only

Formula: C₇H₁₃NO
MW: 127.18 g/mol
MDL Number: MFCD00192246
CAS Number: 13749-61-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR