tert-Butyl-2-methoxypyridin-3-ylcarbamate 98%

Supplier: Ambeed
Warning

A362916-1G A362916-250MG A362916-5G
76828-766EA 53 USD
76828-766 76828-768 77885-957
tert-Butyl-2-methoxypyridin-3-ylcarbamate 98%
tert-Butyl-2-methoxypyridin-3-ylcarbamate

Precaución: for Research Use Only

Formula: C₁₁H₁₆N₂O₃
MDL Number: MFCD11100821
CAS Number: 161117-83-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR