(3,5-Dimethylpyridin-2-yl)methanol 97%

Supplier: Ambeed
Warning

A361577-1G A361577-5G
76828-752EA 28 USD
76828-752 76828-754
(3,5-Dimethylpyridin-2-yl)methanol 97%
(3,5-Dimethylpyridin-2-yl)methanol

Precaución: for Research Use Only

Formula: C₈H₁₁NO
MDL Number: MFCD09839185
CAS Number: 202932-05-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR