2-Bromo-5-(tert-butyl)aniline 98%

Supplier: Ambeed
Warning

Synonyms: 2-Bromo-5-tertbutylaniline

A361261-1G A361261-5G A361261-100G
76828-744EA 20.16 USD
76828-744 76828-746 77594-074
2-Bromo-5-(tert-butyl)aniline 98%
2-Bromo-5-(tert-butyl)aniline

Precaución: for Research Use Only

Formula: C₁₀H₁₄BrN
MW: 228.12 g/mol
MDL Number: MFCD11846050
CAS Number: 58164-14-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR