(3-Methyl-5-propoxyphenyl)boronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3-Methyl-5-propoxyphenylboronic acid

A360359-100MG A360359-1G A360359-250MG
76828-738EA 40 USD
76828-738 76828-740 76828-742
(3-Methyl-5-propoxyphenyl)boronic acid 98%
(3-Methyl-5-propoxyphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₅BO₃
MDL Number: MFCD16295067
CAS Number: 1256346-06-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR