2-(4-Bromo-3-methylphenyl)acetonitrile 98%

Supplier: Ambeed
Warning

A359237-1G A359237-25G A359237-5G
76828-732EA 11 USD
76828-732 76828-734 76828-736
2-(4-Bromo-3-methylphenyl)acetonitrile 98%
2-(4-Bromo-3-methylphenyl)acetonitrile

Precaución: for Research Use Only

Formula: C₉H₈BrN
MDL Number: MFCD14582921
CAS Number: 215800-25-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR