6-Bromo-1,2,3,4-tetrahydronaphthalene 95%

Supplier: Ambeed
Warning

A358556-5G A358556-1G A358556-100MG A358556-250MG
76828-724EA 781.89 USD
76828-724 77568-730 77613-042 77684-055
6-Bromo-1,2,3,4-tetrahydronaphthalene 95%
6-Bromo-1,2,3,4-tetrahydronaphthalene

Precaución: For research use only.

Formula: C₁₀H₁₁Br
MW: 211.10 g/mol
MDL Number: MFCD09842497
CAS Number: 6134-56-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR