N-(3-Acetyl-2-hydroxyphenyl)-4-(4-phenylbutoxy)benzamide 95%, Ambeed.Inc

Supplier: Ambeed
Warning

A358324-10G A358324-1G A358324-250MG A358324-5G A358324-25G
76828-716EA 37.14 USD
76828-716 76828-718 76828-720 76828-722 77684-053
N-(3-Acetyl-2-hydroxyphenyl)-4-(4-phenylbutoxy)benzamide 95%, Ambeed.Inc
N-(3-Acetyl-2-hydroxyphenyl)-4-(4-phenylbutoxy)benzamide

Precaución: For research use only.

Formula: C₂₅H₂₅NO₄
MW: 403.47 g/mol
MDL Number: MFCD09835307
CAS Number: 136450-06-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR