4,5-Bis(di-tert-butylphosphino)-9,9-dimethylxanthene 98%

Supplier: Ambeed
Warning

Synonyms: 9,9-Dimethyl-4,5-bis(di-t-butylphosphino)xanthene, t-Bu-Xantphos, (9,9-Dimethyl-9H-xanthene-4,5-diyl)bis(di-tert-butylphosphine)

A357986-100MG A357986-1G A357986-250MG A357986-5G A357986-25G
76828-706EA 16.98 USD
76828-706 76828-708 76828-710 76828-712 77684-052
4,5-Bis(di-tert-butylphosphino)-9,9-dimethylxanthene 98%
4,5-Bis(di-tert-butylphosphino)-9,9-dimethylxanthene

Precaución: For research use only.

Formula: C₃₁H₄₈OP₂
MW: 498.68 g/mol
MDL Number: MFCD07781995
CAS Number: 856405-77-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR