4-Bromo-3-methyl-2-nitrophenol 98%

Supplier: Ambeed
Danger

Synonyms: 2-Nitro-3-methyl-4-bromophenol

A355416-1G A355416-250MG
76828-674EA 17 USD
76828-674 76828-676
4-Bromo-3-methyl-2-nitrophenol 98%
4-Bromo-3-methyl-2-nitrophenol

Precaución: for Research Use Only

Formula: C₇H₆BrNO₃
MDL Number: MFCD17016113
CAS Number: 85598-12-5
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR