3-Methylpiperidine-2,6-dione 97%

Supplier: Ambeed
Warning

A355058-100MG A355058-1G A355058-250MG
76828-668EA 40 USD
76828-668 76828-670 76828-672
3-Methylpiperidine-2,6-dione 97%
3-Methylpiperidine-2,6-dione

Precaución: for Research Use Only

Formula: C₆H₉NO₂
MDL Number: MFCD13178824
CAS Number: 29553-51-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR