4-Nitro-1H-indazole-6-carboxylic acid 97+%

Supplier: Ambeed
Warning

A354885-100MG A354885-1G A354885-250MG A354885-5G
76828-660EA 25.47 USD
76828-660 76828-662 76828-664 76828-666
4-Nitro-1H-indazole-6-carboxylic acid 97+%
4-Nitro-1H-indazole-6-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₅N₃O₄
MW: 207.14 g/mol
MDL Number: MFCD07781682
CAS Number: 885519-71-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR