7-Bromo-2-methyl-2H-indazole-3-carboxaldehyde 95%

Supplier: Ambeed
Warning

Synonyms: 7-Bromo-2-methyl-2H-indazole-3-carbaldehyde

A354487-100MG A354487-1G A354487-250MG
76828-652EA 76.39 USD
76828-652 76828-654 76828-656
7-Bromo-2-methyl-2H-indazole-3-carboxaldehyde 95%
7-Bromo-2-methyl-2H-indazole-3-carboxaldehyde

Precaución: for Research Use Only

Formula: C₉H₇BrN₂O
MW: 239.08 g/mol
MDL Number: MFCD11520881
CAS Number: 845751-70-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR