2-Bromo-9,9-di-p-tolyl-9H-fluorene 98%

Supplier: Ambeed
Warning

A353837-1G A353837-250MG
76828-648EA 13 USD
76828-648 76828-650
2-Bromo-9,9-di-p-tolyl-9H-fluorene 98%
2-Bromo-9,9-di-p-tolyl-9H-fluorene

Precaución: for Research Use Only

Formula: C₂₇H₂₁Br
MDL Number: MFCD16619315
CAS Number: 474918-33-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR