Ethyl 4-(2-(1,3-dioxoisoindolin-2-yl)ethoxy)-3-oxobutanoate 97%

Supplier: Ambeed
Warning

A353690-100MG A353690-10G A353690-1G A353690-250MG A353690-5G
76828-638EA 29 USD
76828-638 76828-640 76828-642 76828-644 76828-646
Ethyl 4-(2-(1,3-dioxoisoindolin-2-yl)ethoxy)-3-oxobutanoate 97%
Ethyl 4-(2-(1,3-dioxoisoindolin-2-yl)ethoxy)-3-oxobutanoate

Precaución: for Research Use Only

Formula: C₁₆H₁₇NO₆
MDL Number: MFCD12407176
CAS Number: 88150-75-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR