(5-Fluoropyridin-2-yl)methanamine 95%

Supplier: Ambeed
Warning

Synonyms: 2-(Aminomethyl)-5-fluoropyridine

A353092-100MG A353092-1G A353092-250MG A353092-5G
76828-618EA 50.93 USD
76828-618 76828-620 76828-622 76828-624
(5-Fluoropyridin-2-yl)methanamine 95%
(5-Fluoropyridin-2-yl)methanamine

Precaución: for Research Use Only

Formula: C₆H₇FN₂
MW: 126.132 g/mol
MDL Number: MFCD10697659
CAS Number: 561297-96-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR