2-Fluorobenzene-1,4-diamine 95%

Supplier: Ambeed
Danger

A352313-1G A352313-250MG A352313-5G
76828-600EA 109.28 USD
76828-600 76828-602 76828-604
2-Fluorobenzene-1,4-diamine 95%
2-Fluorobenzene-1,4-diamine

Precaución: for Research Use Only

Formula: C₆H₇FN₂
MW: 126.13 g/mol
MDL Number: MFCD09955618
CAS Number: 14791-78-7
UN: 3259
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR