Methyl 4-(1-aminocyclopropyl)benzoate 98%

Supplier: Ambeed
Warning

A350385-1G A350385-250MG
76828-586EA 62 USD
76828-586 77885-921
Methyl 4-(1-aminocyclopropyl)benzoate 98%
Methyl 4-(1-aminocyclopropyl)benzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₃NO₂
MDL Number: MFCD11616340
CAS Number: 1006037-03-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR