3-Bromo-9-ethylcarbazole 95%

Supplier: Ambeed
Danger

Synonyms: 3-Bromo-9-ethyl-9H-carbazole

A350026-1G A350026-250MG
76828-582EA 46.68 USD
76828-582 76828-584
3-Bromo-9-ethylcarbazole 95%
3-Bromo-9-ethylcarbazole

Precaución: for Research Use Only

Formula: C₁₄H₁₂BrN
MW: 274.1558 g/mol
MDL Number: MFCD00092392
CAS Number: 57102-97-3
UN: 1759
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR