7-Chloro-2-methyl-2H-indazole 95%

Supplier: Ambeed
Warning

A349309-100MG A349309-250MG
76828-572EA 79.57 USD
76828-572 76828-574
7-Chloro-2-methyl-2H-indazole 95%
7-Chloro-2-methyl-2H-indazole

Precaución: for Research Use Only

Formula: C₈H₇ClN₂
MW: 166.61 g/mol
MDL Number: MFCD11977512
CAS Number: 1216469-16-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR