(6-(Methoxycarbonyl)pyridin-3-yl)boronic acid 98%, Ambeed.Inc

Supplier: Ambeed
Warning

A348160-10G A348160-250MG A348160-5G A348160-100MG A348160-1G
76828-560EA 378 USD
76828-560 76828-562 76828-564 77684-029 77885-916
(6-(Methoxycarbonyl)pyridin-3-yl)boronic acid 98%, Ambeed.Inc
(6-(Methoxycarbonyl)pyridin-3-yl)boronic acid

Precaución: For research use only.

Formula: C₇H₈BNO₄
MDL Number: MFCD09800559
CAS Number: 1072945-86-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR