Ethyl 4-(cyclopropylamino)butanoate 95%

Supplier: Ambeed
Warning

A346505-100MG A346505-1G A346505-250MG
76828-552EA 42.44 USD
76828-552 76828-554 76828-556
Ethyl 4-(cyclopropylamino)butanoate 95%
Ethyl 4-(cyclopropylamino)butanoate

Precaución: for Research Use Only

Formula: C₉H₁₇NO₂
MW: 171.23 g/mol
MDL Number: MFCD22493469
CAS Number: 813429-65-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR