Hexahydro-1H-furo[3,4-c]pyrrole hydrochloride 95%

Supplier: Ambeed
Warning

A345870-250MG A345870-1G
76828-546EA 183.54 USD
76828-546 77572-006
Hexahydro-1H-furo[3,4-c]pyrrole hydrochloride 95%
Hexahydro-1H-furo[3,4-c]pyrrole hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₂ClNO
MW: 149.61 g/mol
MDL Number: MFCD20527058
CAS Number: 60889-33-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR