(R)-6-(3-Methylpiperazin-1-yl)nicotinonitrile 95%

Supplier: Ambeed
Warning

A344269-1G A344269-250MG
76828-536EA 100 USD
76828-536 76828-538
(R)-6-(3-Methylpiperazin-1-yl)nicotinonitrile 95%
(R)-6-(3-Methylpiperazin-1-yl)nicotinonitrile

Precaución: for Research Use Only

Formula: C₁₁H₁₄N₄
MDL Number: MFCD17488802
CAS Number: 1057682-05-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR