3-Bromo-4-fluorobenzamide 98%
Supplier: Ambeed
|
Warning
|
A343927-25G
A343927-1G
A343927-5G
76828-534EA
201.58
USD
76828-534
77612-966
77684-019
3-Bromo-4-fluorobenzamide 98%
3-Bromo-4-fluorobenzamide
Precaución: For research use only.
Formula:
C₇H₅BrFNO MW: 218.0200 g/mol |
MDL Number:
MFCD03094431 CAS Number: 455-85-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...