4-Benzyloxy-3-methoxyphenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 4-Benzyloxy-3-methoxybenzeneboronic acid, (4-(Benzyloxy)-3-methoxyphenyl)boronic acid

A342437-1G A342437-25G
76828-524EA 35.01 USD
76828-524 76828-526
4-Benzyloxy-3-methoxyphenylboronic acid 98%
4-Benzyloxy-3-methoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₅BO₄
MW: 258.08 g/mol
MDL Number: MFCD09027252
CAS Number: 243990-53-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR