2-Amino-6-chloropyrido[3,2-d]pyrimidin-4(1H)-one 95%

Supplier: Ambeed
Danger

A342188-100MG A342188-1G
76828-518EA 170 USD
76828-518 76828-520
2-Amino-6-chloropyrido[3,2-d]pyrimidin-4(1H)-one 95%
2-Amino-6-chloropyrido[3,2-d]pyrimidin-4(1H)-one

Precaución: for Research Use Only

Formula: C₇H₅ClN₄O
MDL Number: MFCD18373945
CAS Number: 897359-74-9
UN: 3259
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR