(R)-N-(2-Benzoylphenyl)-1-benzylpyrrolidine-2-carboxamide 95%

Supplier: Ambeed
Warning

A341976-1G A341976-250MG A341976-5G
76828-506EA 29 USD
76828-506 76828-508 76828-510
(R)-N-(2-Benzoylphenyl)-1-benzylpyrrolidine-2-carboxamide 95%
(R)-N-(2-Benzoylphenyl)-1-benzylpyrrolidine-2-carboxamide

Precaución: for Research Use Only

Formula: C₂₅H₂₄N₂O₂
MW: 384.48 g/mol
MDL Number: MFCD18910207
CAS Number: 105024-93-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR