1,3-Dibromo-2,2-dimethoxypropane 98%

Supplier: Ambeed
Warning

A341251-100G A341251-10G A341251-1G A341251-5G A341251-25G A341251-500G
76828-488EA 12 USD
76828-488 76828-490 76828-492 76828-494 76839-838 77593-994
1,3-Dibromo-2,2-dimethoxypropane 98%
1,3-Dibromo-2,2-dimethoxypropane

Precaución: for Research Use Only

Formula: BrH₂CC(OCH₃)₂CH₂Br
MDL Number: MFCD00051792
CAS Number: 22094-18-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR