6-Fluoro-4-(trifluoromethyl)quinolin-2(1H)-one 97%

Supplier: Ambeed
Warning

A340935-1G A340935-250MG A340935-5G
76828-478EA 20 USD
76828-478 76828-480 76828-482
6-Fluoro-4-(trifluoromethyl)quinolin-2(1H)-one 97%
6-Fluoro-4-(trifluoromethyl)quinolin-2(1H)-one
Formula: C₁₀H₅F₄NO
MDL Number: MFCD17015723
CAS Number: 328956-08-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR