Methyl-5-bromo-4-chloro-2-fluorobenzoate 98%

Supplier: Ambeed
Danger

Synonyms: Methyl 5-bromo-4-chloro-2-fluorobenzoate

A340076-1G A340076-250MG A340076-5G
76828-472EA 24 USD
76828-472 76828-474 76828-476
Methyl-5-bromo-4-chloro-2-fluorobenzoate 98%
Methyl 5-bromo-4-chloro-2-fluorobenzoate

Precaución: for Research Use Only

Formula: C₈H₅BrClFO₂
MDL Number: MFCD09800902
CAS Number: 951884-02-9
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR