2-(1-Piperidinyl)phenylboronic acid 98%, Ambeed.Inc

Supplier: Ambeed
Warning

Synonyms: (2-(Piperidin-1-yl)phenyl)boronic acid, 2-(Piperidino)phenylboronic acid

A340034-100MG A340034-1G A340034-250MG A340034-5G
76828-466EA 11 USD
76828-466 76828-468 76828-470 77684-010
2-(1-Piperidinyl)phenylboronic acid 98%, Ambeed.Inc
2-(1-Piperidinyl)phenylboronic acid

Precaución: For research use only.

Formula: C₁₁H₁₆BNO₂
MDL Number: MFCD03095111
CAS Number: 1050505-85-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR