Trimethyl-2,2':6',2''-terpyridine-4,4',4''-tricarboxylate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
2,2':6',2''-Terpyridine-4,4',4''-tricarboxylic Acid Trimethyl Ester, Trimethyl [2,2':6',2''-terpyridine]-4,4',4''-tricarboxylate
A339026-100MG
A339026-1G
A339026-250MG
76828-452EA
44.56
USD
76828-452
76828-454
76828-456
Trimethyl-2,2':6',2''-terpyridine-4,4',4''-tricarboxylate 97%
Trimethyl-2,2':6',2''-terpyridine-4,4',4''-tricarboxylate
Precaución: for Research Use Only
Formula:
C₂₁H₁₇N₃O₆ MW: 407.38 g/mol |
MDL Number:
MFCD11042545 CAS Number: 330680-46-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...