Boc-(S)-3-Cyclopentylalanine 95%

Supplier: Ambeed
Warning

Synonyms: (S)-2-((tert-Butoxycarbonyl)amino)-3-cyclopentylpropanoic acid

A338244-100MG A338244-1G A338244-250MG
76828-428EA 45.62 USD
76828-428 76828-430 76828-432
Boc-(S)-3-Cyclopentylalanine 95%
Boc-(S)-3-Cyclopentylalanine

Precaución: for Research Use Only

Formula: C₁₃H₂₃NO₄
MW: 257.326 g/mol
MDL Number: MFCD03094885
CAS Number: 143415-31-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR