4,6-Dichloro-1H-pyrazolo[4,3-c]pyridine 98%

Supplier: Ambeed
Warning

Synonyms: 4,6-Dichloro-5-azaindazole

A337894-100MG A337894-5G
76828-424EA 88.06 USD
76828-424 77550-970
4,6-Dichloro-1H-pyrazolo[4,3-c]pyridine 98%
4,6-Dichloro-1H-pyrazolo[4,3-c]pyridine

Precaución: for Research Use Only

Formula: C₆H₃Cl₂N₃
MW: 188.014 g/mol
MDL Number: MFCD18257114
CAS Number: 1256794-28-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR