Ethyl 3-(4-hydroxyphenyl)acrylate 95+%

Supplier: Ambeed
Warning

A337517-1G A337517-250MG A337517-5G
76828-418EA 32 USD
76828-418 76828-420 76828-422
Ethyl 3-(4-hydroxyphenyl)acrylate 95+%
Ethyl 3-(4-hydroxyphenyl)acrylate

Precaución: for Research Use Only

Formula: C₁₁H₁₂O₃
MDL Number: MFCD00210460
CAS Number: 2979-06-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR